Assay Details
| Gene Symbol: Dnajc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109180.1 | NP_001102650.1 | 4 - 5 | 595 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P898347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157563 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P898347 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P898347 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P898347 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P898347 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50129 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814411 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814412 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814409 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200005150.rn.1 | TC0200000850.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005151.rn.1 | TC0200000850.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005154.rn.1 | TC0200000850.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005152.rn.1 | TC0200000850.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10814409 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22358327 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366204 | - | - |

Dnajc5b
Genomic Map