Assay Details
| Gene Symbol: Egfem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107663.1 | NP_001101133.1 | 3 - 4 | 613 | 93 | |
| XM_008760923.1 | XP_008759145.1 | 3 - 4 | 701 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Egfem1
Genomic Map