Assay Details
| Gene Symbol: Lgals7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228668.2 | XP_006228730.1 | 3 - 4 | 270 | 106 | |
| XM_006228670.2 | XP_006228732.1 | 2 - 3 | 442 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228668.2, XM_006228670.2 transcripts.
Set Membership: |
|

Lgals7
Genomic Map