Assay Details
| Gene Symbol: Lgals7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022582.2 | NP_072104.2 | 2 - 3 | 297 | 103 | |
| XM_006228668.2 | XP_006228730.1 | 5 - 6 | 555 | 103 | ||
| XM_006228670.2 | XP_006228732.1 | 4 - 5 | 727 | 103 | ||
| GenBank mRNA | AF036941.1 | - | 2 - 3 | 306 | 103 | |
| BI278774.1 | - | 1 - 2 | 103 | |||
| U67883.1 | - | 3 - 4 | 306 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals7
Genomic Map