Assay Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008352.1 | NP_001008353.1 | 3 - 4 | 364 | 154 | |
| GenBank mRNA | BC086349.1 | - | 3 - 4 | 364 | 7302798 | 154 |
| FQ220630.1 | - | 3 - 4 | 414 | 154 | ||
| FQ226470.1 | - | 3 - 4 | 494 | 154 | ||
| FQ233923.1 | - | 3 - 4 | 494 | 154 | ||
| FQ234368.1 | - | 3 - 4 | 494 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmvk
Genomic Map