Assay Details
| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079711.1 | NP_001073179.1 | 7 - 8 | 907 | 107 | |
| XM_006237502.2 | XP_006237564.1 | 7 - 8 | 890 | 107 | ||
| XM_006237504.2 | XP_006237566.1 | 7 - 8 | 904 | 107 | ||
| GenBank mRNA | BC128747.1 | - | 7 - 8 | 907 | 7099220 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|

Rad51ap1
Genomic Map