Assay Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037285.1 | NP_001032362.1 | 2 - 3 | 217 | 107 | |
| NM_001037286.1 | NP_001032363.1 | 1 - 2 | 259 | 107 | ||
| NM_001037287.1 | NP_001032364.1 | 2 - 3 | 174 | 107 | ||
| NM_022397.3 | NP_071792.1 | 2 - 3 | 188 | 107 | ||
| GenBank mRNA | BC097275.1 | - | 2 - 3 | 157 | 7444983 | 107 |
| BC103634.1 | - | 2 - 3 | 176 | 7097633 | 107 | |
| CK359545.1 | - | 1 - 2 | 259 | 107 | ||
| CK366752.1 | - | 2 - 3 | 174 | 107 | ||
| CK473366.1 | - | 2 - 3 | 248 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097275.1, BC103634.1, NM_001037285.1, NM_001037286.1, NM_001037287.1, NM_022397.3 transcripts.
Set Membership: |
|

Hnrnpf
Genomic Map