Assay Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037285.1 | NP_001032362.1 | 1 - 2 | 77 | 114 | |
| GenBank mRNA | CK473366.1 | - | 1 - 2 | 108 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001037285.1 transcripts.
Set Membership: |
|

Hnrnpf
Genomic Map