Assay Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008314.1 | NP_001008315.1 | 5 - 6 | 859 | 147 | |
| GenBank mRNA | BC083870.1 | - | 5 - 6 | 859 | 7128406 | 147 |
More Information
Updated Mapping Information
Set Membership: |
|

Mkrn2
Genomic Map