Assay Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008314.1 | NP_001008315.1 | 6 - 7 | 977 | 159 | |
| GenBank mRNA | BC083870.1 | - | 6 - 7 | 977 | 7128406 | 159 |
More Information
Updated Mapping Information
Set Membership: |
|

Mkrn2
Genomic Map