Assay Details
| Gene Symbol: Fam21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199207.1 | NP_954677.1 | 1 - 2 | 78 | 128 | |
| XM_006237147.2 | XP_006237209.1 | 1 - 2 | 102 | 128 | ||
| XM_006237149.2 | XP_006237211.1 | 1 - 2 | 102 | 128 | ||
| GenBank mRNA | AY257251.1 | - | 1 - 2 | 78 | 128 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY257251.1,NM_199207.1,XM_006237147.2,XM_006237149.2 transcripts.
Set Membership: |
|

Fam21c
Genomic Map