Assay Details
| Gene Symbol: Fam21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199207.1 | NP_954677.1 | 24 - 25 | 2678 | 81 | |
| XM_006237147.2 | XP_006237209.1 | 24 - 25 | 2702 | 81 | ||
| XM_006237149.2 | XP_006237211.1 | 23 - 24 | 2600 | 81 | ||
| GenBank mRNA | AY257251.1 | - | 24 - 25 | 2678 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21c
Genomic Map