Assay Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012745.2 | NP_036877.1 | 5 - 6 | 669 | 82 | |
| GenBank mRNA | AF009133.1 | - | 5 - 6 | 426 | 82 | |
| BC086393.1 | - | 5 - 6 | 669 | 7303354 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klrd1
Genomic Map