Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031649.2 | NP_113837.1 | 2 - 3 | 185 | 89 | |
| GenBank mRNA | X79812.1 | - | 2 - 3 | 238 | 89 | |
| Gene Symbol: RGD1561215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_575659.5 | XP_575659.3 | 1 - 2 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klrg1
Genomic Map