Assay Details
| Gene Symbol: Zfp384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034830.1 | NP_001030002.1 | 9 - 10 | 1384 | 78 | |
| NM_001034831.1 | NP_001030003.1 | 8 - 9 | 1200 | 78 | ||
| NM_133429.2 | NP_596920.2 | 9 - 10 | 1467 | 78 | ||
| XM_006237326.2 | XP_006237388.1 | 7 - 8 | 1245 | 78 | ||
| XM_006237327.2 | XP_006237389.1 | 10 - 11 | 1432 | 78 | ||
| XM_006237328.2 | XP_006237390.1 | 7 - 8 | 1180 | 78 | ||
| XM_006237329.2 | XP_006237391.1 | 7 - 8 | 1110 | 78 | ||
| GenBank mRNA | AB019281.1 | - | 9 - 10 | 1384 | 78 | |
| AF216804.1 | - | 9 - 10 | 1467 | 78 | ||
| AF216805.1 | - | 7 - 8 | 1186 | 78 | ||
| AF216806.1 | - | 8 - 9 | 1200 | 78 | ||
| AF216807.1 | - | 10 - 11 | 2132 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp384
Genomic Map