Assay Details
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173291.1 | NP_775413.1 | 1 - 2 | 105 | 120 | |
| GenBank mRNA | AY649838.1 | - | 1 - 2 | 116 | 120 | |
| U56822.1 | - | 1 - 2 | 105 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY649838.1, NM_173291.1, U56822.1 transcripts.
Set Membership: |
|

Klra22
Genomic Map