Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134645.1 | NP_001128117.1 | 9 - 10 | 1517 | 58 | |
| XM_006246701.2 | XP_006246763.1 | 10 - 11 | 1891 | 58 | ||
| XM_006246702.2 | XP_006246764.1 | 9 - 10 | 2165 | 58 | ||
| XM_008767825.1 | XP_008766047.1 | 9 - 10 | 1225 | 58 | ||
| GenBank mRNA | EF532342.1 | - | 9 - 10 | 1517 | 58 | |
| EF532343.1 | - | 9 - 10 | 1569 | 58 | ||
| EF532344.1 | - | 9 - 10 | 1576 | 58 | ||
| EF532345.1 | - | 9 - 10 | 1569 | 58 | ||
| EF532346.1 | - | 9 - 10 | 1576 | 58 | ||
| EF532347.1 | - | 9 - 10 | 1576 | 58 | ||
| EF532348.1 | - | 9 - 10 | 1576 | 58 | ||
| EF532349.1 | - | 9 - 10 | 1576 | 58 | ||
| EF532350.1 | - | 1576 | 58 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cntrob
Genomic Map