Assay Details
| Gene Symbol: Rnmtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108282.1 | NP_001101752.1 | 1 - 2 | 325 | 82 | |
| GenBank mRNA | FQ232324.1 | - | 1 - 2 | 323 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnmtl1
Genomic Map