Assay Details
| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253859.1 | NP_001240788.1 | 5 - 6 | 491 | 59 | |
| XM_008767967.1 | XP_008766189.1 | 4 - 5 | 418 | 59 | ||
| GenBank mRNA | BC169112.1 | - | 5 - 6 | 489 | 59 | |
| CK596986.1 | - | 5 - 6 | 504 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tom1l1
Genomic Map