Assay Details
| Gene Symbol: Ppm1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105825.2 | NP_001099295.2 | 5 - 6 | 1454 | 68 | |
| GenBank mRNA | BC162058.1 | - | 5 - 6 | 1454 | 7115026 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P672273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168239 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P672273 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P672273 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P672273 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19411 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10737070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737072 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737071 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737074 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737069 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737073 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10737068 | - | - |
| Rat Genome 230 2.0 Array | 1374884_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374884_at | - | - |
| Affymetrix HT Rat Focus Array | 1374884_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374884_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000011936.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011937.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011939.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011941.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011942.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011943.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011933.rn.1 | TC1000001298.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000011938.rn.1 | TC1000001298.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374884_at | - | - |
| Rat Expression Set 230 | 1374884_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737068 | - | - |
| Rat Genome U34 Set | rc_AI233712_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21331864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365461 | - | - |

Ppm1d
Genomic Map