Assay Details
| Gene Symbol: Ikzf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107047.1 | NP_001100517.1 | 7 - 8 | 881 | 85 | |
| XM_008768049.1 | XP_008766271.1 | 2 - 3 | 978 | 85 | ||
| XM_008768050.1 | XP_008766272.1 | 2 - 3 | 852 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
regulation of B cell proliferation
regulation of apoptotic process
regulation of B cell differentiation
regulation of lymphocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity
transcription from RNA polymerase II promoter
regulation of B cell proliferation
regulation of apoptotic process
regulation of B cell differentiation
regulation of lymphocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P088261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107432 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088261 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088261 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P088261 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P424635 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23448 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10746913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746907 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10746905 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000033220.rn.1 | TC1000003716.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10746905 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20796894 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372439 | - | - |

Ikzf3
Genomic Map