Assay Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183327.1 | NP_899156.1 | 3 - 4 | 318 | 78 | |
| XM_006246125.2 | XP_006246187.1 | 3 - 4 | 881 | 78 | ||
| GenBank mRNA | AY574252.1 | - | 3 - 4 | 318 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrg2
Genomic Map