Assay Details
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102364.1 | NP_001095834.1 | 1 - 2 | 112 | 59 | |
XM_008767511.1 | XP_008765733.1 | 2 - 3 | 343 | 59 | ||
GenBank mRNA | BC168250.1 | - | 1 - 2 | 112 | 5598970 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168250.1,NM_001102364.1,XM_008767511.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
