Assay Details
| Gene Symbol: LOC102549389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768230.1 | XP_008766452.1 | 2 - 3 | 233 | 72 | |
| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012914.1 | NP_037046.1 | 5 - 6 | 595 | 72 | |
| XM_006246597.2 | XP_006246659.1 | 6 - 7 | 1209 | 72 | ||
| GenBank mRNA | M30581.1 | - | 5 - 6 | 595 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102549389
Genomic Map