Assay Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017960.1 | NP_001017960.1 | 5 - 6 | 801 | 90 | |
| NM_031134.2 | NP_112396.2 | 4 - 5 | 669 | 90 | ||
| GenBank mRNA | CB728734.1 | - | 1 - 2 | 150 | 90 | |
| M18028.1 | - | 5 - 6 | 691 | 90 | ||
| M31174.1 | - | 883 | 90 | |||
| X07409.1 | - | 3 - 4 | 330 | 90 | ||
| X12744.1 | - | 4 - 5 | 333 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thra
Genomic Map