Assay Details
| Gene Symbol: Rai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271207.1 | NP_001258136.1 | 4 - 5 | 5960 | 79 | |
| GenBank mRNA | AY724505.1 | - | 3 - 4 | 800 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rai1
Genomic Map