Assay Details
| Gene Symbol: LOC103690023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069291.5 | XP_001069291.2 | 2 - 3 | 408 | 85 | |
| XM_008773812.1 | XP_008772034.1 | 1 - 2 | 696 | 85 | ||
| Gene Symbol: LOC689039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768152.1 | XP_008766374.1 | 1 - 2 | 696 | 85 | |
| XM_008768153.1 | XP_008766375.1 | 2 - 3 | 408 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690023
Genomic Map