Assay Details
| Gene Symbol: Pkmyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105766.1 | NP_001099236.1 | 6 - 7 | 1638 | 104 | |
| XM_006245868.2 | XP_006245930.1 | 6 - 7 | 1184 | 104 | ||
| GenBank mRNA | BC169008.1 | - | 5 - 6 | 1731 | 7387434 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Pkmyt1
Genomic Map