Assay Details
| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271189.1 | NP_001258118.1 | 2 - 3 | 301 | 96 | |
| GenBank mRNA | FQ227491.1 | - | 2 - 3 | 312 | 96 | |
| FQ233678.1 | - | 2 - 3 | 301 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c
Genomic Map