Assay Details
| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271189.1 | NP_001258118.1 | 4 - 5 | 476 | 97 | |
| GenBank mRNA | FQ227491.1 | - | 4 - 5 | 487 | 97 | |
| FQ233678.1 | - | 4 - 5 | 476 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c
Genomic Map