Assay Details
| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009625.1 | NP_001009625.1 | 1 - 2 | 161 | 61 | |
| GenBank mRNA | BC088222.1 | - | 1 - 2 | 161 | 7321956 | 61 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC088222.1,NM_001009625.1 transcripts.
Set Membership: |
|

Ifi35
Genomic Map