Assay Details
| Gene Symbol: Ifi35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009625.1 | NP_001009625.1 | 5 - 6 | 708 | 70 | |
| GenBank mRNA | BC088222.1 | - | 5 - 6 | 708 | 7321956 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Ifi35
Genomic Map