Assay Details
| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037773.4 | NP_001032862.2 | 2 - 3 | 272 | 65 | |
| XM_006246133.2 | XP_006246195.1 | 3 - 4 | 958 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnjl
Genomic Map