Assay Details
| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004200.1 | NP_001004200.1 | 10 - 11 | 1184 | 64 | |
| GenBank mRNA | BC079143.1 | - | 10 - 11 | 1184 | 7128107 | 64 |
| FQ213575.1 | - | 6 - 7 | 718 | 64 | ||
| FQ213666.1 | - | 6 - 7 | 715 | 64 | ||
| FQ228882.1 | - | 10 - 11 | 1225 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cops3
Genomic Map