Assay Details
| Gene Symbol: Ormdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247345.1 | XP_006247407.1 | 1 - 2 | 497 | 163 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247345.1 transcripts.
Set Membership: |
|

Ormdl3
Genomic Map