Assay Details
| Gene Symbol: LOC103690164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246210.2 | XP_006246272.1 | 7 - 8 | 1060 | 81 | |
| XM_008767733.1 | XP_008765955.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767734.1 | XP_008765956.1 | 1890 | 81 | |||
| XM_008767735.1 | XP_008765957.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767736.1 | XP_008765958.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767737.1 | XP_008765959.1 | 1890 | 81 | |||
| XM_008767738.1 | XP_008765960.1 | 9 - 10 | 1890 | 81 | ||
| Gene Symbol: Phykpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128196.1 | NP_001121668.1 | 9 - 10 | 1092 | 81 | |
| XM_008767645.1 | XP_008765867.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767647.1 | XP_008765869.1 | 9 - 10 | 1153 | 81 | ||
| XM_008767648.1 | XP_008765870.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767649.1 | XP_008765871.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767650.1 | XP_008765872.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767651.1 | XP_008765873.1 | 9 - 10 | 1890 | 81 | ||
| XM_008767652.1 | XP_008765874.1 | 9 - 10 | 1890 | 81 | ||
| GenBank mRNA | BC079163.1 | - | 9 - 10 | 1105 | 81 | |
| BC158766.1 | - | 9 - 10 | 1092 | 7114211 | 81 | |
| FQ209628.1 | - | 1 - 2 | 121 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690164
Genomic Map