Assay Details
| Gene Symbol: Skp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007608.2 | NP_001007609.1 | 2 - 3 | 179 | 113 | |
| XM_006246244.2 | XP_006246306.1 | 2 - 3 | 478 | 113 | ||
| GenBank mRNA | BC058152.1 | - | 2 - 3 | 182 | 6890712 | 113 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Skp1
Genomic Map