Assay Details
| Gene Symbol: Ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199489.4 | NP_955783.1 | 2 - 3 | 139 | 110 | |
| GenBank mRNA | AY379972.1 | - | 2 - 3 | 142 | 110 | |
| BC089762.1 | - | 126 | 7381637 | 110 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY379972.1,BC089762.1,NM_199489.4 transcripts.
Set Membership: |
|

Ccr7
Genomic Map