Assay Details
| Gene Symbol: Rnf167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008361.1 | NP_001008362.1 | 2 - 3 | 777 | 74 | |
| XM_006246750.2 | XP_006246812.1 | 2 - 3 | 191 | 74 | ||
| GenBank mRNA | BC083670.1 | - | 2 - 3 | 777 | 7135651 | 74 |
| FQ216790.1 | - | 2 - 3 | 111 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf167
Genomic Map