Assay Details
| Gene Symbol: Enox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770914.1 | XP_008769136.1 | 12 - 13 | 2638 | 68 | |
| XM_008770915.1 | XP_008769137.1 | 12 - 13 | 2635 | 68 | ||
| XM_008770916.1 | XP_008769138.1 | 11 - 12 | 2581 | 68 | ||
| GenBank mRNA | FQ222793.1 | - | 2263 | 68 | ||
More Information
Set Membership: |
|

Enox1
Genomic Map