Assay Details
| Gene Symbol: Nudt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100732.1 | NP_001094202.1 | 2 - 3 | 455 | 81 | |
| GenBank mRNA | BC082050.1 | - | 388 | 81 | ||
| BC098710.1 | - | 2 - 3 | 457 | 81 | ||
| BC158874.1 | - | 2 - 3 | 455 | 7933460 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nudt18
Genomic Map