Assay Details
| Gene Symbol: Med4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024256.1 | NP_001019427.1 | 4 - 5 | 436 | 159 | |
| GenBank mRNA | BC093402.1 | - | 4 - 5 | 436 | 7389622 | 159 |
| FQ231796.1 | - | 2 - 3 | 122 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med4
Genomic Map