Assay Details
| Gene Symbol: Atp6v0e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053578.3 | NP_446030.3 | 1 - 2 | 198 | 72 | |
| GenBank mRNA | AB037248.1 | - | 1 - 2 | 198 | 72 | |
| FQ224739.1 | - | 1 - 2 | 159 | 72 | ||
| FQ226644.1 | - | 1 - 2 | 161 | 72 | ||
| FQ226841.1 | - | 1 - 2 | 168 | 72 | ||
| FQ233078.1 | - | 1 - 2 | 157 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp6v0e1
Genomic Map