Assay Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012010.1 | NP_001012010.1 | 2 - 3 | 535 | 107 | |
| GenBank mRNA | AY539935.1 | - | 494 | 107 | ||
| BC087718.1 | - | 2 - 3 | 535 | 7316530 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf135
Genomic Map