Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044224.1 | NP_001037689.1 | 15 - 16 | 2725 | 87 | |
| GenBank mRNA | AB231661.1 | - | 15 - 16 | 2725 | 87 | |
| AF111111.1 | - | 3 - 4 | 297 | 87 | ||
| BC166408.1 | - | 15 - 16 | 2715 | 9030944 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag5
Genomic Map