Assay Details
| Gene Symbol: Krt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173128.1 | NP_775151.1 | 5 - 6 | 1161 | 67 | |
| GenBank mRNA | M63665.1 | - | 5 - 6 | 1161 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt20
Genomic Map