Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105809.2 | NP_001099279.1 | 3 - 4 | 292 | 64 | |
| GenBank mRNA | BC166808.1 | - | 3 - 4 | 292 | 7124384 | 64 |
| FQ228687.1 | - | 3 - 4 | 308 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dph1
Genomic Map