Assay Details
| Gene Symbol: Smg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105808.1 | NP_001099278.1 | 12 - 13 | 3733 | 60 | |
| XM_008767960.1 | XP_008766182.1 | 2 - 3 | 202 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smg6
Genomic Map