Assay Details
| Gene Symbol: LOC103694863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773805.1 | XP_008772027.1 | 4 - 5 | 2505 | 70 | |
| XM_008773806.1 | XP_008772028.1 | 4 - 5 | 1089 | 70 | ||
| XM_008773807.1 | XP_008772029.1 | 5 - 6 | 1309 | 70 | ||
| XM_008773808.1 | XP_008772030.1 | 4 - 5 | 1306 | 70 | ||
| XM_008773809.1 | XP_008772031.1 | 3 - 4 | 993 | 70 | ||
| XM_008773810.1 | XP_008772032.1 | 4 - 5 | 1072 | 70 | ||
| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191648.1 | NP_001178577.1 | 3 - 4 | 527 | 70 | |
| XM_006247028.2 | XP_006247090.1 | 4 - 5 | 1089 | 70 | ||
| XM_006247029.2 | XP_006247091.1 | 5 - 6 | 1309 | 70 | ||
| XM_006247030.2 | XP_006247092.1 | 4 - 5 | 1306 | 70 | ||
| XM_008768033.1 | XP_008766255.1 | 4 - 5 | 2505 | 70 | ||
| XM_008768034.1 | XP_008766256.1 | 4 - 5 | 1072 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694863
Genomic Map