Assay Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105764.1 | NP_001099234.1 | 1 - 2 | 275 | 127 | |
XM_006245862.2 | XP_006245924.1 | 1 - 2 | 292 | 127 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105764.1,XM_006245862.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
